Hydrochloric acid 6.0 N, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1,06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:5.95 - 6.05
- Normality:5.95 - 6.05
Specification Test Results
Frequently Bought Together