

Nonidet® P 40 Substitute (NP-40), Proteomics Grade
Catalog # M158-100ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Density (20 °C):1.05 - 1.07 g/ml
- Protease:NONE
- Refractive Index:1.48 - 1.52
- Solubility (5%, Water):PASS
Specification Test Results
About this item
:
Boiling Pt:290 °C (1013 hPa)
Melting Pt:57…58 °C
Density:1,06 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-024-6
UN:3082
ADR:9,III
:GHS07
:GHS09
CAS Number:9016-45-9
MDL Number:MFCD00132851