Stopcock keys for burettes, DURAN®
Catalog # SCOT286035504
Supplier: DWK LIFE SCIENCES
Specifications
- For:Burette stopcock
- Hole Ø:2,5 mm
- Description:Stopcock keys, side entry, drilled and ground, with three part retaining device
- Pk:1
- Spigot:12,5 NS
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
- Side entry
- Drilled and grounded
- With three part retaining device