VWR® RiboZol™, RNA Extraction Reagents
Supplier: VWR Chemicals
Certificates
About this item
Ribozol™ RNA Extraction Reagent is a single phase phenol solution optimized for the isolation of total RNA from a variety of cell and tissue types.
- Glass bottle
Homogenization or disruption directly in Ribozol™ RNA Extraction Reagent inhibits RNase activity to substantially minimize degradation of all classes of RNA. The simple and effective procedure for isolation in Ribozol™ RNA Extraction Reagent includes homogenization, phase separation, RNA precipitation, RNA wash and solubilization.
Specifications
- Method / Format:Organic extraction
- Prep size:Scalable
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:RiboZol™ RNA Extraction Reagents
Specifications
Frequently Bought Together