


Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99.0%, Biotechnology Grade for biotechnology 1 Phase
Catalog # K169-400ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:400 mL
- Seq:2665
Specifications
Specification Test Results
- Conductivity(50%, Water) @25 °C:250 uS - 330 uS
- pH(50%, Water) @25°C:8.05 - 8.35
Specification Test Results
About this item
:
UN:2810
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:136112-00-0
MDL Number:MFCD00133763