Order Entry
India
ContactUsLinkComponent
Sodium chlorite, high purity
Sodium chlorite, high purity
Catalog # N819-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chlorite, high purity
Catalog # N819-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Arsenic:
    <3 ppm
  • Heavy Metals (as Pb):
    <20 ppm
  • Hydrogen Peroxide:
    0.01 %
  • Lead:
    <10 ppm
  • Mercury:
    <0.1 ppm
  • Sodium Carbonate:
    <2 %
  • Sodium Chlorate:
    <4%
  • Sodium Chloride:
    11 – 19 %
  • Sodium Chlorite:
    77.5 – 82.5 %
  • Sodium Hydroxide:
    <3%
  • Sodium Sulphate:
    <3%
  • Water (by difference):
    <6%

Specification Test Results

About this item

 :
 :90.44 g/mol
Merck Index:13,08672
Boiling Pt:112 °C (1013 hPa)
Melting Pt:250…257 °C
Density:1,28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-836-6
UN:1908
ADR:8,II
 :GHS03
 :GHS06
 :GHS05
 :GHS09
CAS Number:7758-19-2
Formula:ClNaO₂
MDL Number:MFCD00003478