Order Entry
India
ContactUsLinkComponent
 
 
 
 
 
Cyclohexane ≥99% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
  V552-10
 :  
undefined
 
 
 
 
Cyclohexane ≥99% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
  V552-10
 :  

 

  • Pk:
    4 L
  • Seq:
    5162

 

 

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • ACS - Assay (C₆H₁₂) (by GC, corrected for water):
    ≥ 99.0 %
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Residue after Evaporation:
    ≤ 2 ppm
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤ 0.01 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:
    ≤ 1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤ 0.60
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤ 0.25
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤ 0.15
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤ 0.08
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:
    ≤ 0.03
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤ 0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 - 400 nm:
    ≤ 0.01
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤ 1.0 ppb
  • ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):
    ≤ 10 ppb
  • Trace Impurities - GC-FID Sensitive Impurities (as 2-Octanol) Single Peak (µg/L):
    ≤ 10 ppb

 

 

 :
 :84.16 g/mol
 :13,02752
 :81 °C (1013 hPa)
 :7 °C
 :0,7785 g/cm³ (20 °C)
 :–18 °C
 :Ambient
 :203-806-2
 :1145
 :3,II
 :Management of Change (MOC) category = NI
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-82-7
 :C₆H₁₂
 :MFCD00003814