Toluene ≥99.8% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
V560-10
:
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- Environmentally Preferable:
- ACS - Assay (C₆H₅CH₃) (by GC, corrected for water):≥99.8 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0002 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Water (H₂O)(by KF, coulometric):≤0.02 %
- ACS - Sulfur Compounds (as S):≤0.003 %
- Optical Absorbance (1-cm path vs water) - 285 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 286 nm:≤0.70
- Optical Absorbance (1-cm path vs water) - 288 nm:≤0.40
- Optical Absorbance (1-cm path vs water) - 293 nm:≤0.20
- Optical Absorbance (1-cm path vs water) - 300 nm:≤0.10
- Optical Absorbance (1-cm path vs water) - 310 nm:≤0.05
- Optical Absorbance (1-cm path vs water) - 335 nm:≤0.02
- Optical Absorbance (1-cm path vs water) - 400 - 350 nm:≤0.01
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):≤10
: Methylbenzene, Phenylmethane, FM approval
:
:92.14 g/mol
:13,09607
:110,6 °C (1013 hPa)
:–95 °C
:0,867 g/cm³ (20 °C)
:4 °C
:Ambient
:203-625-9
:1294
:3,II
:05-2114615130-69
:GHS02
:GHS08
:GHS07
:108-88-3
:C₆H₅CH₃
:MFCD00008512