Acetone ≥99.5%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56,2 °C (1013 hPa)
Melting Pt:–95,4 °C
Density:0,792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- ACS - Assay (CH₃COCH₃) (by GC, corrected for water):≥99.5 %
- ACS - Aldehydes (as HCHO):≤0.002 %
- ACS - Color (APHA):≤10
- ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- ACS - Methanol (CH₃OH)(by GC):≤0.05 %
- ACS - Residue after Evaporation:≤0.0001 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Titrable Base (meq/g):≤0.0006
- ACS - Water (H₂O):≤0.5 %
- Optical Absorbance (1-cm path vs water) - 330 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 340 nm:≤0.06
- Optical Absorbance (1-cm path vs water) - 400 - 350 nm:≤0.01
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):≤10 ppb
- FID Sensitive Impurities (as 2-Octano l) Single Impurity Peak (ng/mL):≤5 ppb
Specification Test Results
Frequently Bought Together