Order Entry
India
Orders LinkContactUsLinkComponent
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations, 4 L
Catalog # BDHB3492-4
Supplier:   BDH US PRODUCED
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # BDHB3492-4
Supplier:  BDH US PRODUCED
CAS Number:  
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations, 4 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 4 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    4 L
  • Description:
    TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

Specifications