

Trichloroethylene ≥99.5% (by GC, corrected for water content) stabilised, AR® ACS, Macron Fine Chemicals™
MACR8600-04
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- ACS - Assay (ClCH:CCl₂) (by GC, corrected for water):≥99.5 %
- ACS - Titrable Acid (meq/g):≤0.0001
- ACS - Titrable Base (meq/g):≤0.0003
- ACS - Free Halogens:Passes Test
- ACS - Heavy Metals (as Pb):≤0.00001 %
- ACS - Residue after Evaporation:≤0.0005 %
- ACS - Water (H₂O)(by coulometry):≤0.02 %
- ACS - Color (APHA):≤10
:
:12,09769
:86,7 °C (1013 hPa)
:–73 °C
:1,46 g/cm³ (20 °C)
:6.1,III
:Management of Change (MOC) category = N
:131.39 g/mol
:Ambient
:201-167-4
:1710
:GHS08
:GHS07
:79-01-6
:ClCH=CCl₂
:MFCD00000838