Transfection reagent, TransIT®-LT1
Supplier: MIRUS
Certificates
About this item
The broad spectrum, ready-to-use TransIT ®-LT1 transfection reagent is based on histone polyamine and offers high transfection efficiency, low cell toxicity, and is suitable for a wide variety of cell types. Reagents for RNA and electroporation, and cell line specific reagents are also available, please enquire for details.
Specifications
- DNA:Yes
- Virus production:Yes