Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Photometric test kits for Nickel
Photometric test kits for Nickel
Photometric test kits for Nickel
Catalog # 705-0106
Supplier:
HACH
CAS Number:
Certificates
Expand image
Photometric test kits for Nickel
Catalog # 705-0106
Supplier:
HACH
CAS Number:
Certificates
Specifications
Chemical method:
PAN
Pk:
100 Tests
Description:
Nickel
Sample size:
25 ml
Type:
RS
For unit:
DR 2800, DR 4000, DR 5000
Range:
0,007 - 1,000 Ni mg/l
Specifications
About this item
Product Details
Documentation
Product Details
Documentation