Order Entry
India
ContactUsLinkComponent
Manganese(II) sulphate monohydrate 98.0-101.0%, powder, AR® ACS, Macron Fine Chemicals™
Health
Environment
Manganese(II) sulphate monohydrate 98.0-101.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR6192-20
CAS Number:  
undefined
Health
Environment
Manganese(II) sulphate monohydrate 98.0-101.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR6192-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (MnSO₄·H₂O):
    98.0 - 101.0 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.002 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.002 %
  • ACS - Loss on Ignition:
    10.0 - 12.0 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Nickel (Ni):
    ≤0.02 %
  • ACS - Potassium (K):
    ≤0.01 %
  • ACS - Sodium (Na):
    ≤0.05 %
  • Solubility:
    Passes Test
  • ACS - Substances Reducing KMnO₄:
    Passes Test
  • ACS - Zinc (Zn):
    ≤0.005 %

Specification Test Results

About this item

 :
Merck Index:13,05763
Melting Pt:700 °C
Density:2,95 g/cm³ (25 °C)
Storage Temperature:Refrigerator
ADR:9,III
 :169.02 g/mol
EINECS:232-089-9
 :GHS08
 :GHS09
CAS Number:10034-96-5
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159