Order Entry
India
ContactUsLinkComponent
 
 
 
Sodium metabisulphite ≥97%, GPR RECTAPUR®
 
undefined
 
 
Sodium metabisulphite ≥97%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 97 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

 

Frequently Bought Together