


Potassium permanganate ≥99% ACS
Catalog # 0277-2.5KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bucket
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Chloride & Chlorate:<0.005%
- Insolubles:≤0.2%
- Purity:≥99%
- Sulphate:≤0.02%
Specification Test Results
About this item
:
:158.03 g/mol
Merck Index:12,07824
Melting Pt:> 240 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-760-3
UN:1490
ADR:5.1,II
:GHS03
:GHS07
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364