Stopcock, single way, DURAN®
Supplier: DWK LIFE SCIENCES
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
- Very good chemical resistance
- Very good temperature resistance
Straight bore.
Frequently Bought Together