Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
Density Meters
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Catalog # GRAND16
Supplier:
GRANT INSTRUMENTS
Expand image
Accessories for Densitometer for Cell Suspensions
Catalog # GRAND16
Supplier:
GRANT INSTRUMENTS
Specifications
Description:
Spare adaptor for Ø 16 mm tubes
Seq:
901
Pk:
1
Specifications
About this item
Product Details
Product Details