


1-Butanol ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:71-36-3
:
:H₃C(CH₂)₂CH₂OH
:MFCD00002964
:74.12 g/mol
:13,01539
:117,6 °C (1013 hPa)
:–89 °C
:0,812 g/cm³ (20 °C)
:30 °C
:Ambient
:200-751-6
:1120
:3,III
:01-2119484630-38
:GHS02
:GHS07
:GHS05
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Assay (on anhydrous substance):Min. 99.5 %
- IR Spectrum:Passes test
- Acidity or alkalinity:Max. 0.0005 meq/g
- Boiling point:117 - 119 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.807 - 0.812
- Density (20/20):0.808 - 0.813
- Aldehydes + ketones (as C3H7CHO):Max. 0.01 %
- Butan-2-ol:Max. 0.05 %
- Dibutyl ether:Max. 0.2 %
- Evaporation residue:Max. 10 ppm
- Water:Max. 0.1 %
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Frequently Bought Together