


Diethyl ether ≥99.0% stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9259-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay ((C₂H₅)₂O) (by GC, exclusive of preservative, corrected for water):≥99.0 %
- Diethyl Ether Interferences - Suitability for U.S. EPA Method 8151A:Passes Test
- Color (APHA):≤10 ppm
- Residue after Evaporation:≤1.0 ppm
- Peroxide (as H₂O₂):≤5 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Titrable Acid (µeq/g):≤0.2
- Water (by KF, coulometric):≤0.08 %
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0,7135 g/cm³ (20 °C)
Flash Pt:–45 °C
ADR:3,I
:Packaged under inert gas.
:Stabilised with ethanol
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
:GHS02
:GHS07
:GHS08
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646