Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Perlite
Perlite for high clarity filtration
Perlite for high clarity filtration
:
VWR Chemicals
Perlite for high clarity filtration
:
VWR Chemicals
:
93763-70-3
:
:
> 1093 °C
:
2,2…2.4 g/cm³ (20 °C)
pH (10 %):
6.50 - 7.50
Permeability (Darcy):
1.0 - 2.3
Density (g/l):
192 - 232
30L Plastic bucket UN Z (ZPACK13247033)
5L Plastic bucket UN Z (ZPACK13247005)
sds_VWRC84101
spec_VWRC84101.290
spec_VWRC84101.360
VWR® for Drying and Filtration
VWR® Green Solvents for Sustainable Chemistry.pdf
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Bottles, HDPE
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Phosphate 1,000 mg/L in Water standard for ion chromatography
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals