Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7558-80-7
:
Formula:NaH₂PO₄
MDL Number:MFCD00003527
Merck Index:12,08806
Melting Pt:∼650 °C
Density:0,9996 g/cm³ (20 °C)
:119.98 g/mol
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Heavy Metals (as Pb):0.001%
- Insolubles:= 0.03%
- Moisture (KF):= 0.3%
- pH (1%, Water) @25°C:4.4 - 4.7
- Purity:>= 98%
Specification Test Results
Frequently Bought Together