Lanthanum(III) chloride heptahydrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10025-84-0
:
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756
Boiling Pt:< 860 °C (1013 hPa)
Melting Pt:96 °C
:371.37 g/mol
Storage Temperature:Ambient
EINECS:233-237-5
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Frequently Bought Together