Order Entry
India
ContactUsLinkComponent
 
Tin(II) chloride dihydrate ≥95%, TECHNICAL
  23738.232
 :  
undefined
Tin(II) chloride dihydrate ≥95%, TECHNICAL
  23738.232
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Assay:
    Min. 95 %

 

 

 :
 :225.65 g/mol
 :13,08861
 :623 °C (1013 hPa)
 :246 °C
 :2,71 g/cm³ (25 °C)
 :Ambient
 :231-868-0
 :3260
 :8,III
 :GHS07
 :GHS05
 :10025-69-1
 :SnCl₂·2H₂O
 :MFCD00149863