Sulphuric acid 10%, AnalaR NORMAPUR® analytical reagent
Catalog # 9755.1000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:10 - 11 %
- Appearance:Clear colourless liquid
- Substances reducing KMnO4 (as SO2):Passes test
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- NO3 (Nitrate):Max. 10 ppm
- As (Arsenic):Max. 3 ppm
- Fe (Iron):Max. 0.02 %
- Pb (Lead):Max. 5 ppm
- Se (Selenium):Max. 20 ppm
Specification Test Results
About this item
:
:98.08 g/mol
Boiling Pt:330
EINECS:231-639-5
UN:3264
:GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.066 g/cm³ (20 °C)
ADR:8 III