Hydrofluoric acid 48 - 51%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
9563-01
: Avantor
:
- Pack type:PE bottle
- Pk:500 mL
- Seq:2725
- For Laboratory,Research,or Manufacturing Use:48.0 - 51.0 %
- Assay (HF) (by acidimetry):48.0 - 51.0 %
- Color (APHA):≤10
- Residue after Ignition:≤5 ppm
- Fluosilicic Acid (H₂SiF₆):≤0.010 %
- Chloride (Cl):≤5 ppm
- Nitrate (NO₃):≤3 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate and Sulfite (as SO₄):≤5 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤30.0 ppb
- Trace Impurities - Barium (Ba):≤50.0 ppb
- Trace Impurities - Beryllium (Be):≤20.0 ppb
- Trace Impurities - Bismuth (Bi):≤100.0 ppb
- Trace Impurities - Boron (B):≤50 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤50.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤5.0 ppb
- Trace Impurities - Copper (Cu):≤5.0 ppb
- Trace Impurities - Gallium (Ga):≤20.0 ppb
- Trace Impurities - Germanium (Ge):≤50.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Heavy Metals (as Pb):≤100 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤20.0 ppb
- Trace Impurities - Lithium (Li):≤20.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤50.0 ppb
- Trace Impurities - Mercury (Hg):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤50.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silver (Ag):≤20.0 ppb
- Trace Impurities - Sodium (Na):≤200.0 ppb
- Trace Impurities - Strontium (Sr):≤20.0 ppb
- Trace Impurities - Tantalum (Ta):≤100.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤50.0 ppb
- Trace Impurities - Vanadium (V):≤50.0 ppb
- Trace Impurities - Zinc (Zn):≤20.0 ppb
- Trace Impurities - Zirconium (Zr):≤20.0 ppb
:
:20.01 g/mol
:Ambient
:231-634-8
:1790
:8,II
:GHS06
:GHS05
:7664-39-3
:HF
:MFCD00011346