Order Entry
India
ContactUsLinkComponent
Hydrochloric acid 25% Reag. Ph. Eur.
Hydrochloric acid 25% Reag. Ph. Eur.
Catalog # 84514.360
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid 25% Reag. Ph. Eur.
Catalog # 84514.360
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 25.0 %

Specification Test Results

About this item

Synonyms: Hydrochloric acid solution, Muriatic acid
 :
 :36.46 g/mol
EINECS:231-595-7
UN:1789
REACH:01-2119484862-27
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Melting Pt:–30 °C
Density:1.13 g/cm³ (20 °C)
ADR:8 II