Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Dimethylbenzene
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0,86 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
REACH:01-2119488216-32
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (mix of 3 isomers + ethylbenzene):Min. 98.5 %
- IR Spectrum:Passes test
- Acidity or alkalinity:Max. 0.0002 meq/g
- Boiling point:137 - 141 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.865 - 0.867
- Density (20/20):0.866 - 0.869
- Refractive index (20°C):1.495 - 1.499
- Substances coloured by H2SO4:Max. 1600 APHA
- Benzene:Max. 0.02 %
- Ethylbenzene:Max. 25 %
- Evaporation residue:Max. 10 ppm
- Sulphur compounds (as S):Max. 5 ppm
- Thiophene:Max. 1 ppm
- Toluene:Max. 1.0 %
- Water:Max. 0.03 %
- Al (Aluminium):Max. 0.05 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.05 ppm
- Ca (Calcium):Max. 0.2 ppm
- Cd (Cadmium):Max. 0.02 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.05 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.05 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.02 ppm
- Sn (Tin):Max. 0.05 ppm
- Zn (Zinc):Max. 0.05 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
28975_Xylene_BSE_rev0
BSE/TSE Certificates
spec_VWRC28975.291 spec_VWRC28975.291 spec_VWRC28975.325 spec_VWRC28975.360 spec_VWRC28975.462
Product specification
More
Frequently Bought Together