Bovine serum albumin (BSA), Fraction V
Catalog # A2244.0050
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- CAS No.:90604-29-8
9048-46-8 - Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- pH:pH 5.2