Accessories for OLS200 and GLS Aqua Plus shaking water baths
Supplier: GRANT INSTRUMENTS
Certificates
About this item
A range of different stainless steel accessories for use with OLS and GLS Aqua Plus shaking water baths.
- Universal flask tray secures various size containers from 25 ml to 1000 ml
- Tray for test tube racks, holds up to five test tube racks
- Test tube racks in various formats, these are perforated to aid convection
- Perforated tray enables shaking baths to be used as unstirred baths
- Sloping lids direct condensation down the side of the bath, protecting vessels in the bath. PC lids are supplied as standard with GLS Aqua Plus and OLS200 models
Caution: * Test tube racks are used with the appropriate stainless steel tray, 3 per tray (GLS Aqua 12 Plus) or 5 (OLS200 and GLS Aqua 18 Plus).
** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
Frequently Bought Together