Order Entry
India
ContactUsLinkComponent
Magnetic hotplate stirrer, MR Hei-Tec
Magnetic hotplate stirrer, MR Hei-Tec
Catalog # HEID505-30080-06
undefined
Magnetic hotplate stirrer, MR Hei-Tec
Catalog # HEID505-30080-06
Specifications
  • Description:
    MR Silver magnetic hotplate stirrer package includes MR Hei-Tec magnetic hotplate stirrer with IN-plug, PT 1000 probe and probe fixing system
  • Seq:
    08
  • Pk:
    1
Specifications
About this item
Magnetic hotplate stirrer with electronic temperature control and digital display for nominal speed and temperature. Ceramic coated silumin plate provides excellent heat conduction and distribution and is extremely resistant to scratches and chemicals. An independent safety circuit switches off the heating if the nominal hotplate temperature is exceeded by 25 °C. Electronic current limiting to protect the motor. TRIAC guarantees long service life.
  • Digital display for speed and temperature
  • Extra safety control circuit and hotplate cut-out by two independent temperature sensors
  • Connector for electronic temperature control (via optional PT1000 probe)
  • Medium temperature range: Up to 250 °C, temperature control accuracy with optional PT1000 probe: ±1 °C
IP protection class according to DIN EN 60529: IP 32.