Order Entry
India
ContactUsLinkComponent
Deep well plates, 96- and 48- well, Axygen®
Deep well plates, 96- and 48- well, Axygen®
  738-0188
 :  Corning
   Low Manufacturing Impact
undefined
Deep well plates, 96- and 48- well, Axygen®
  738-0188
 :  Corning
   Low Manufacturing Impact
  • Description:
    Deep 96-well plate, conical wells, 0,6 ml, sterile, individually wrapped
  • Seq:
    16
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    25
PP. Used for sample collection, cell culture and long term storage.
  • Excellent chemical resistance and temperature resistance down to −80 °C
  • Can withstand centrifugation up to 4000×g
Two types of sealing mats are suitable for use with these plates:
AxyMat™ sealing mats are designed for high throughput screening and storage applications. Made of silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be re-used. AxyMat™ sealing mats are pierceable and self-sealing, ideal for automated processes, and can be used in sub-zero environments.
ImpermaMat™ sealing mats are chemically resistant. They are made for use in assays involving strong solvents such as DMSO and similarly harsh solvents. Re-usable. They feature a tight seal to the plate so there is no sample evaporation or well-to-well evaporation. Usable within a wide temperature range: from −80 up to +121 °C.
ImpermaMat™ sealing mats are chemically resistant.  They are made for use in assays involving strong solvents such as DMSO and similarly harsh solvents.  Re-usable.  They feature a tight seal to the plate so there is no sample evaporation or well-to-well evaporation. Usable within a wide temperature range: from −80 °C up to +121 °C.