Order Entry
India
ContactUsLinkComponent
Industrial gloves, JACKSON SAFETY* G40 Nitrile Coated
Industrial gloves, JACKSON SAFETY* G40 Nitrile Coated
Catalog # 112-2575
Supplier:  KIMBERLY CLARK
undefined
Industrial gloves, JACKSON SAFETY* G40 Nitrile Coated
Catalog # 112-2575
Supplier:  KIMBERLY CLARK
Specifications
  • Glove size:
    11
  • Ambidextrous / hand-specific:
    Hand-specific
  • Anti-static:
    +
  • Disposable / reusable:
    Reusable
  • Colour:
    Black/blue
  • Length:
    220 - 260 mm
  • Seq:
    05
  • Model:
    Nitrile Coated
  • EN norm:
    EN 388 (4131), EN 1149-2
  • Cuff style:
    Knit wrist
  • Liner material:
    Nylon
  • Gauge:
    15
  • Finish:
    Palm and fingertips coated
  • Pk:
    12 PAIR
Specifications
About this item
Nitrile foam coated hand specific gloves with seamless knitted nylon liner.
  • Excellent dexterity
  • Abrasion resistance rating of 4 (EN Standards)
  • Excellent wet and dry grip
  • Static dissipative in use
  • Silicone-free
  • Ecofriendly packaging - no separate insert, no printed bags, less waste
Ideal for applications where tactile sensitivity is required, for example in metal manufacturing and automotive and aerospace assembly.
Certifications: EN 388 (4131), EN 1149-2; Cat. II