Order Entry
India
ContactUsLinkComponent
VWR®, Flash Chromatography Cartridges
Catalog # 554-0265
Supplier:  VWR Collection
undefined
VWR®, Flash Chromatography Cartridges
Catalog # 554-0265
Supplier:  VWR Collection
Specifications
  • Column Format:
    Flash cartridge
  • Description:
    Empty cartridge with seals, frits and filters
  • Seq:
    28
  • Pk:
    25
Specifications
About this item
Resistant to all normal phase solvents used.
  • Allow for liquid or solid sample loading
  • Axial compression of silica: no dead volume
Components for MiniVarioFlash®, internal Ø 12 mm
Product Information