Bottles, wide neck, square for solid products
Catalog # 13251000.
Supplier: VWR Chemicals
Specifications
- Thread:DIN 60
- Capacity:1000 ml
- W×D×H:97×97×210 mm
- Neck I-Ø:50,5 mm
- Pk:1
Specifications
About this item
PE.
- Ideal for storing and shipping samples
- Wide mouth permits easy filling, emptying and cleaning
- Square shape saves space