Order Entry
India
ContactUsLinkComponent
 
Methyl cellulose, 4000 mPa.S (2% H₂O), high purity
  K390-250G
 :  
undefined
Methyl cellulose, 4000 mPa.S (2% H₂O), high purity
  K390-250G
 :  

 

  • Pack type:
    Bottle, amber
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility (2%, Water):
    Pass
  • Viscosity (2%, water) @20 °C:
    3500 – 5600 cps
  • Appearance:
    white powder with a yellow cast

 

 

 :
 :Ambient
 :9004-67-5
 :MFCD00081763