MultiScreen™ PH Plates
PH plates, 0,28 cm² filtration area, 28L×85,5W×14,4H mm, 0,65 μm pore size, phosphocellulose membrane
Catalog # MILYMAPHN0B50
Supplier: MILLIPORE
Jump to:
Product Details & Documents
Reliable tools for the advancement of drug discovery and life science research.
- Negatively charged phosphocellulose
- Low protein/nucleic acid binding
- Opaque
- Non sterile
- Volume: 0,05 to 0,25 ml
Application: Protein kinase assays with peptide substrates, enzyme assays, binds positive charge.
Specifications
- Description:PH plates, 0,28 cm² filtration area, 28L×85,5W×14,4H mm, 0,65 μm pore size, phosphocellulose membrane



