


Quinaldine red 0,1% in methanol Reag. Ph. Eur. 1073801
Catalog # 87923.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:430.33 g/mol
Storage Temperature:Ambient
EINECS:204-221-5
UN:1230
ADR:3 (6.1),II
:GHS02
:GHS06
:GHS08
CAS Number:117-92-0
Formula:C₂₁H₂₃IN₂
MDL Number:MFCD00011968