Order Entry
India
ContactUsLinkComponent
Potassium Ferriperiodate Solution Reag. Ph. Eur. 1070801
Potassium Ferriperiodate Solution Reag. Ph. Eur. 1070801
Catalog # 87917.130
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium Ferriperiodate Solution Reag. Ph. Eur. 1070801
Catalog # 87917.130
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
UN:1719
ADR:8,II