Order Entry
India
ContactUsLinkComponent
Potassium heptaiodobismuthate 6,1% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601
Potassium heptaiodobismuthate 6,1% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601
Catalog # 87913.270
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium heptaiodobismuthate 6,1% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601
Catalog # 87913.270
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    600 mL
  • Seq:
    2775

Specifications

About this item

 :
 :1253.71 g/mol
EINECS:255-595-1
CAS Number:41944-01-8
Formula:BiI₇K₄