Order Entry
India
ContactUsLinkComponent
Ninhydrin and stannous chloride reagent R1 (a+b)
Ninhydrin and stannous chloride reagent R1 (a+b)
Catalog # 87876.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ninhydrin and stannous chloride reagent R1 (a+b)
Catalog # 87876.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 SET
  • Seq:
    0125
  • Pk Info:
    2x100 ml

Specifications

About this item

 :
Storage Temperature:Ambient
UN:1188
ADR:3,III