Order Entry
India
ContactUsLinkComponent
VWR® Class II, Type B2 EN 12469 Biosafety Cabinet with Integrated Particle Monitoring System (IPMS)
VWR® Class II, Type B2 EN 12469 Biosafety Cabinet with Integrated Particle Monitoring System (IPMS)
Supplier:  VWR Collection
Certificates
undefined
VWR® Class II, Type B2 EN 12469 Biosafety Cabinet with Integrated Particle Monitoring System (IPMS)
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 135-2898
  • 135-2896
  • 135-2897
  • 135-2895
  • Model:
  • VWRA135-2898
  • VWRA135-2896
  • VWRA135-2897
  • VWRA135-2895
  • Filters:
  • 2 HEPA H14 Efficiency 99.9995% at 0.3 um, DOP tested
  • 2 HEPA H14 Efficiency 99.9995% at 0.3 um, DOP tested
  • 2 HEPA H14 Efficiency 99.9995% at 0.3 um, DOP tested
  • 2 HEPA H14 Efficiency 99.9995% at 0.3 um, DOP tested
  • Lighting system:
  • Eco-friendly LED lighting average >1000 lux/chamber red light alarm
  • Eco-friendly LED lighting average >1000 lux/chamber red light alarm
  • Eco-friendly LED lighting average >1000 lux/chamber red light alarm
  • Eco-friendly LED lighting average >1000 lux/chamber red light alarm
  • Sash opening:
  • 450 mm
  • 450 mm
  • 450 mm
  • 450 mm
  • Ext. W×D×H:
  • 1830×800×1450 mm
  • 1220×800×1450 mm
  • 1525×800×1450 mm
  • 915×800×1450 mm
  • Int. W×D×H:
  • 1715×600×640 mm
  • 1135×600×640 mm
  • 1440×600×640 mm
  • 835×600×640 mm

Specifications

Related Information

Frequently Bought Together