Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fuchsin solution, decolourised
Fuchsin solution, decolourised R1 R1 Reag. Ph. Eur. 1039402
Fuchsin solution, decolourised R1 R1 Reag. Ph. Eur. 1039402
Fuchsin solution, decolourised R1 R1 Reag. Ph. Eur. 1039402
Catalog # 87840.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Fuchsin solution, decolourised R1 R1 Reag. Ph. Eur. 1039402
Catalog # 87840.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
UN:
1789
ADR:
8,III
Product Details
Documentation