Nitro blue tetrazolium chloride (NBT) ≥98.0% for molecular biology
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:298-83-9
:
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159
:817.65 g/mol
Melting Pt:200 °C
EINECS:206-067-4
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Assay:Min. 98.0 %
- Appearance:Yellow powder
- IR Spectrum:Passes test
- Water:Max. 5.0 %
- Sulphated ash:Max. 0.20 %
- Absorptivity (256.6 nm):Passes test
- Control by TLC:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC438592X spec_VWRC438597T spec_VWRC438597T spec_VWRC438597T
Product specification
Frequently Bought Together