VWR® LED Trigger Box for Colibri 5 and 7
Catalog # 612-7306
Supplier: VWR Collection
New Product
Specifications
- Description:LED trigger box for Colibri 5/7
- Pk:1
- For:Optional, for triggering with 3rd party hardware
Specifications
About this item
LED trigger box for Colibri 5/7 for fast control of the illumination module via TTL and analog signals from external software packages is needed.