Order Entry
India
ContactUsLinkComponent
 
Dichloromethane ≥99.50%, Laboratory Reagent, Rankem®
 :  RANKEM
 
Dichloromethane ≥99.50%, Laboratory Reagent, Rankem®
 :  RANKEM
 

 

  • BIS Specification:
    .
  • Purity:
    ≥ 99.50 %
  • Relative Density at 27/ 27°C:
    1.314 - 1.319
  • Residue On Evaporation:
    ≤ 0.001 %
  • Moisture:
    ≤ 0.02 %
  • Acidity (as HCl):
    ≤ 0.0003 %
  • Free Chlorine:
    Passes test
  • Aldehydes:
    Passes test
  • Iron (Fe):
    Max. 0.04 ppm
  • Heavy Metals (As Pb):
    ≤ 1.00 ppm
  • Resistance to potassium permanganate:
    Passes test
  • Nessler’s reagent:
    Passes test
  • Resistance to sulphuric acid:
    Passes test
  • Total impurities due to methyl chloride and chloroform:
    ≤ 0.10 %
  • LR Specifications:
    .
  • Weight Per ml (at 20°C):
    1.323 - 1.326 g
  • Identity (IR):
    Conforms
  • Colour:
    ≤ 10 APHA
  • Amylene (As Stabilizer):
    30 - 150 ppm

 

Frequently Bought Together