

Trichloroethylene ≥99.00%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:79-01-6
:
Formula:ClCH=CCl₂
MDL Number:MFCD00000838
Boiling Pt:86,7 °C (1013 hPa)
Melting Pt:–73 °C
EINECS:201-167-4
UN:1710
ADR:6.1,III
:131.39 g/mol
:GHS07
:GHS08
Specification Test Results
- Assay:Min. 99.00%
- Weight Per ml (At 20 °C):1.462 --> 1.466 g
- Distillation Range:86.0 - 87.5 °C
- Acidity (As HCl):Max. 0.01000%
- Free Chlorine:≤ 0.001%
- Non-Volatile Matter:Max. 0.00400%
- Water (By KF):Max. 0.050%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_RANKT0180 spec_RANKT0200 spec_RANKT0205 spec_RANKT0250
Product specification
Frequently Bought Together