Order Entry
India
ContactUsLinkComponent
Ferritin Rapid Quantitative Test
Ferritin Rapid Quantitative Test
  FINCW269P0001
 :  FINECARE
 :  
Ferritin Rapid Quantitative Test
  FINCW269P0001
 :  FINECARE
 :  
  • Pk:
    1
  • Description:
    Ferritin rapid quantitative test
  • Sample type:
    Serum/plasma/whole blood
  • Processing time:
    10 min
  • Range:
    5 - 1000 ng/ml
The Finecare™ Ferritin Rapid Quantitative Test is a fluorescence immunoassay used along with Finecare™ FIA Meters for quantitative determination of Ferritin in human whole blood, serum or plasma. The test is used as an aid to the assessment of iron deficiency anemia.
  • Store the test kit at 4 to 30 ℃ up to the expiration date printed on the package
  • Do not remove the test cartridge from the pouch until ready to use
  • The test cartridge should be used within 1 hour once opened