cTnT Rapid Quantitative Test
cTnT rapid quantitative test, 1
Catalog # FINCW254P0001
Supplier: FINECARE
Jump to:
Product Details & Documents
Internationally recognised gold standard for AMI diagnosis. Good sensitivity and specificity for early myocardial infraction.
- Supports whole blood/serum/plasma sample type
- Room temperature storage up to 24 months
- One step operation, streamline the workflow
Diagnose: AHA/ACC: Cardiac specific troponin levels should be measured at presentation and 3 to 6 hours after symptom onset. Monitoring: Change value from serial troponin tests for better monitoring and making decision. Admission (0 h), 3, and 6 h serial Troponin test are preferred for clinicians in ED. Prognosis: An elevated troponin level can identify patients at high risk. Higher mortality is correlating with the degree of troponin elevation in ICU patients.
Specifications
Specifications
- Pk:1
- Description:cTnT rapid quantitative test
- Cat. No.:FINCW254P0001
- Sample type:Serum/plasma/whole blood
- Size:75 µl serum/plasma or 100 µl whole blood
- Processing time:15 mins
- Shelf life:4...30 °C for 24 months
- Range:0,03 - 10 ng/ml



