

Chloroform ≥99.0%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):99.00 --> 99.40%
- Weight Per ml (at 20 °C):1.475 --> 1.481 g
- Color:≤ 10 APHA
- Stabilizer:0.600 --> 1.000%w/w
- Maximum Limits Of Impurities:.
- Residue On Evaporation:Max. 0.0010%
- Water ( By KF):Max. 0.100%
Specification Test Results
Related Information
Frequently Bought Together